missing translation for 'onlineSavingsMsg'
Learn More

Uridine 5'-monophosphate, disodium salt, hydrate, 98%

Catalog No. AC226290050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226290050 5 g Glass bottle
AC226290010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226290050 Supplier Thermo Scientific Chemicals Supplier No. 226290050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

homogeneous | CAS: 3387-36-8 | C9H11N2Na2O9P | 368.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3387-36-8
Molecular Formula C9H11N2Na2O9P
Molecular Weight (g/mol) 368.15
MDL Number MFCD00149422
InChI Key KURVIXMFFSNONZ-IJRYJGAHNA-L
Synonym uridine-5'-monophosphate disodium salt, uridine 5'-monophosphate, disodium salt, 5'-uridylicaciddisodiumsalt
PubChem CID 123134060
SMILES [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=CC(=O)NC1=O

Specifications

Absorbance 0.71 to 0.77 (A250/A260),0.38 to 0.44 (A280/A260)
Melting Point 208.0°C to 210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 10, 9689
Solubility Information Solubility in water:+/- 40% (20°C)
Spectrophotometry 97.5% min. (lambda max. 262nm,pH 7) molar absorptivity 10000+/-2%,on dry basis
Transmittance 95% min. (c=1, 430nm) 1cm cell
IUPAC Name [(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
Formula Weight 368.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Uridine 5′-monophosphate, disodium salt, hydrate
Show More Show Less
EINECSNumber 222-211-9

RUO – Research Use Only