missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Uridine-5'-monophosphate disodium salt, 99%

Catalog No. AAA1860106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1860106 5 g
AAA1860114 25 g
AAA1860122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1860106 Supplier Thermo Scientific Chemicals Supplier No. A1860106
Only null left

CAS: 3387-36-8 | C9H11N2Na2O9P | 368.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3387-36-8
Molecular Formula C9H11N2Na2O9P
Molecular Weight (g/mol) 368.15
MDL Number MFCD00149422
InChI Key KURVIXMFFSNONZ-IJRYJGAHNA-L
Synonym uridine-5'-monophosphate disodium salt, uridine 5'-monophosphate, disodium salt, 5'-uridylicaciddisodiumsalt
PubChem CID 123134060
IUPAC Name [(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
SMILES [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=CC(=O)NC1=O

Specifications

Melting Point ∼209°C (decomposition)
Quantity 5 g
Beilstein 3582885
Formula Weight 368.15
Percent Purity 99%
Chemical Name or Material Uridine-5'-monophosphate disodium salt
EINECSNumber 222-211-9
RTECSNumber YU7975000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only