missing translation for 'onlineSavingsMsg'
Learn More

Uricase, Candida utilis, Thermo Scientific Chemicals

Catalog No. aaj60875exr
Change view
Click to view available options
Quantity:
100 U
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ60875EXR 100 U Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60875EXR Supplier Thermo Scientific Chemicals Supplier No. J60875EXR
Only null left

 

Uricase is very important for the determination of uric acid in biological fluids. Uricase is an important medical enzyme which can be used to determine urate in clinical analysis. The enzyme Urate oxidase (UO) catalyzes the oxidation of uric acid to 5-hydroxyisourate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12-4-9002
Molecular Formula (C3H4O2)n
Molecular Weight (g/mol) 72.06
MDL Number MFCD00084394
InChI Key NNMHYFLPFNGQFZ-UHFFFAOYSA-M
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
PubChem CID 6581
ChEBI CHEBI:18308
IUPAC Name prop-2-enoic acid
SMILES OC(=O)C(-*)C-*

Specifications

Packaging Plastic bottle
Quantity 100 U
Solubility Information Hardly soluble in water,slightly soluble in buffer alkaline solution.
Physical Form Lyophilized Powder
Chemical Name or Material Uricase
EINECSNumber 232-655-5
RTECSNumber YU7100000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only