missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PureCoat™ Carboxyl Rectangular Straight Neck Cell Culture Flask with Vented Cap
Description
Corning™ PureCoat™ amine (positively charged) and carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
Specifications
| Closure Material | HDPE |
| Capacity (Metric) | 513 mL |
| Material | Polystyrene |
| Neck Style | Straight |
| Product Type | Cell Culture Flask |
| Shape | Rectangular |
| Surface Coating | Carboxyl |
| Disposable | Single-use |
| Graduated | Yes |
| Sterility | Sterile |
| Show More |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.