missing translation for 'onlineSavingsMsg'
Learn More

Urease from Jack Bean, TCI America™

$240.46 - $1275.02

Chemical Identifiers

CAS 9002-13-5
MDL Number MFCD00070858
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
U0017100MG
View Documents
TCI America
U0017100MG
100 mg
Each for $240.46
Only null left
 
U00171G
View Documents
TCI America
U00171G
1 g
Each for $1,275.02
Only null left
 
Description

Description

Specifications

Chemical Identifiers

9002-13-5
MFCD00070858

Specifications

9002-13-5
MFCD00070858
Crystalline Powder
White-Yellow
100 mg
Urease from Jack Bean
Videos
Safety and Handling

Safety and Handling

RTECSNumber : YU1700000

TSCA : Yes

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'