missing translation for 'onlineSavingsMsg'
Learn More

Urea (USP), Fisher Chemical™

Catalog No. U163
Change view
Click to view available options
Quantity:
3 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
U163 3 kg Poly Bottle
U1650 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U163 Supplier Thermo Fisher Scientific Supplier No. U16 3
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Decomposition Information Decomposes information: >132°C
Melting Point 131°C
Color White
Density 1.335g/cm³
pH 7.5 to 9.5
Ignition Residue 0.1% max. (USP)
Melting Range 132° to 135°C (USP)
Packaging Poly Bottle
Linear Formula NH2CONH2
Quantity 3 kg
Solubility Information 0.04% max. (USP) Insoluble in Alcohol
Residual Solvents Meets Requirements (USP)
Vapor Pressure 1.25mmHg at 25°C
Formula Weight 60.06
Percent Purity 99.0 to 100.5% (USP)
Grade USP
Physical Form Solid
Chemical Name or Material Urea
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation.
NFPA
Health:1 Flammability:0 Instability:0