missing translation for 'onlineSavingsMsg'
Learn More

Urea, ultrapure, 99%

Catalog No. p-7043149
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6576922 100 g
AAJ6576936 500 g
AAJ65769A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6576922 Supplier Thermo Scientific Chemicals Supplier No. J6576922
Only null left

CAS: 57-13-6 | CH4N2O | 60.056 g/mol

Urea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Urea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.

Solubility
Soluble in waterSoluble in water, alcohol, methanol, glycerol, benzene and concentrated hydrochloric acid. Practically insoluble in chloroform and ether.

Notes
Stable at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 132°C to 135°C
Color White
Density 1.335
pH 9.2 to 9.5
Odor Ammonia-like
Linear Formula NH2CONH2
Quantity 100 g
Beilstein 635724
Merck Index 14,9867
Solubility Information Soluble in waterSoluble in water,alcohol,methanol,glycerol,benzene and concentrated hydrochloric acid; Practically insoluble in chloroform and ether.
Formula Weight 60.06
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Urea
Grade Ultrapure
Show More Show Less
EINECSNumber 200-315-5
RTECSNumber YR6250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only