missing translation for 'onlineSavingsMsg'
Learn More

Urea, Ultra Pure, 5lb, MP Biomedicals

Catalog No. ICN821527
Change view
Click to view available options
Quantity:
5 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN821527 5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN821527 Supplier MP Biomedicals Inc Supplier No. 04821527
Only null left

57-13-6 Purity: ≥99% Heavy metals (as Pb): <<0.002 ppm O.D. (260nm) of a 7M solution: <<0.05 Protein denaturant

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Quantity 5 lb.
Grade Ultrapure
Chemical Name or Material Urea