missing translation for 'onlineSavingsMsg'
Learn More

Urea, Shotted Pellets, Reagents

Catalog No. p-200021142
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336899 2.5 kg
01336898 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336899 Supplier Labchem Inc Supplier No. C23455002.5A5
Only null left

Urea, Shotted Pellets, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
IUPAC Name urea
SMILES NC(N)=O

Specifications

Boiling Range 135°C
Melting Point 133°C
Color White
Boiling Point 135°C
Melting Range 133°C
Packaging Poly Bottle
Quantity 2.5 kg
Specific Gravity 1.335
Grade Technical
Physical Form Solid
Chemical Name or Material Urea, Shotted Pellets
Show More Show Less