missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Urea, OmniPur™, Calbiochem™,

Catalog No. 9510500GM
Change view
Click to view available options
Quantity:
500 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9510500GM 500 g
95305KG 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 9510500GM Supplier MilliporeSigma™ Supplier No. 9510500GM

Ideal for protein unfolding/denaturation

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 132.8°C
Color White
pH 10.1 (25°C)
Quantity 500 g
Solubility Information Water: Soluble
Formula Weight 60.07
Concentration or Composition (by Analyte or Components) 0.9 to 1.1M
Physical Form Solid
Chemical Name or Material Urea
Grade Molecular biology grade
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.