missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Urea, Molecular biology grade, Calbiochem™,

Catalog No. 666122500GM
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Alu Drum
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
666122500GM 500 g Glass Bottle
66612225KG 2.5 kg Alu Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 666122500GM Supplier MilliporeSigma™ Supplier No. 666122500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym Carbamide
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Color White
Quantity 500 g
Packaging Glass Bottle
Percent Purity ≥98%
Grade Molecular Biology
Chemical Name or Material Urea