missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Urea, Molecular biology grade, Calbiochem™,

Supplier:  MilliporeSigma™ 66612225KG

Quantity 2.5 kg
Packaging Alu Drum

 View more versions of this product

Catalog No. 66612225KG


Only null left

Description

Description

Specifications

Chemical Identifiers

57-13-6
60.056
Carbamide
CHEBI:48376
C(=O)(N)N
CH4N2O
XSQUKJJJFZCRTK-UHFFFAOYSA-N
1176
urea

Specifications

White
2.5 kg
CH4N2O
XSQUKJJJFZCRTK-UHFFFAOYSA-N
urea
1176
≥98%
57-13-6
Alu Drum
Carbamide
C(=O)(N)N
60.056
CHEBI:48376
Molecular Biology
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions