missing translation for 'onlineSavingsMsg'
Learn More

Urea, (Crystalline/Reagent Grade), MP Biomedicals

Supplier:  MP Biomedicals Inc 0210320905

Catalog No. ICN10320905

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Urea
CH4N2O
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
C(=O)(N)N
60.056
CHEBI:48376
Reagent
57-13-6
5 kg
XSQUKJJJFZCRTK-UHFFFAOYSA-N
urea
1176
60.1
Crystalline

Chemical Identifiers

57-13-6
60.056
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
CHEBI:48376
C(=O)(N)N
CH4N2O
XSQUKJJJFZCRTK-UHFFFAOYSA-N
1176
urea
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions