missing translation for 'onlineSavingsMsg'
Learn More

Urea (Certified ACS), Fisher Chemical™

Catalog No. U153
Change view
Click to view available options
Quantity:
500 g
3 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
U153 3 kg Poly Bottle
U15500 500 g Poly Bottle
U1550 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. U153 Supplier Thermo Fisher Scientific Supplier No. FLU153
Only null left
Edge

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Decomposition Information Decomposes information: >132°C
Melting Point 131°C
Color White
Density 1.335g/cm³
pH 7.5 to 9.5
Ignition Residue 0.01% max.
Assay Percent Range 99.0 to 100.5 %
Packaging Poly Bottle
Linear Formula NH2CONH2
Quantity 3 kg
Solubility Information 0.01% max. Insoluble Matter
Vapor Pressure 1.25mmHg at 25°C
Formula Weight 60.06
Percent Purity 99.0 to 100.5%
Grade Certified ACS
Physical Form Solid
Chemical Name or Material Urea
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation.
NFPA
Health:1 Flammability:0 Instability:0