missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Urea, Calbiochem™,

Catalog No. 666121KG
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
666121KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 666121KG Supplier MilliporeSigma™ Supplier No. 666121KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity crystalline protein denaturant used to increase the solubility of hydrophobic molecules.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym Carbimide
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Color White
Quantity 1 kg
Packaging Glass Bottle
Percent Purity ≥98%
Chemical Name or Material Urea