Learn More
Bioassay Systems QuantiChrom™ Urea Assay Kit. For quantitative determination of urea and evaluation of drug effects on urea metabolism.
Supplier:  Bioassay Systems DIUR100
Encompass
For quantitative determination of urea and evaluation of drug effects on urea metabolism. Key Features: Sensitive and accurate. Use 5 µL samples. Linear detection range 0.08 mg/dL (13 µM) to 100 mg/dL (17 mM) urea in 96-well plate assay. Simple and high-throughput. Can be readily automated as a high-throughput assay for thousands of samples per day. Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Cuvet or 96-well plate assay. Low interference in biological samples. No pretreatments are needed. Assays can be directly performed on raw biological samples i.e., in the presence of lipid and protein. Method: OD520nm (Chemical). Samples: Serum, plasma, urine, milk, cell/tissue culture, bronchoalveolar lavage (BAL), food, beverage and environment. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.08 mg/dL (13 µM).
Catalog No. 501078333