missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea, ACS Reagent, Reagents
Specifications
Specifications
| Boiling Range | 135°C |
| Melting Point | 133°C |
| Color | White |
| Boiling Point | 135°C |
| Melting Range | 133°C |
| Packaging | Glass Bottle |
| Quantity | 100 g |
| Specific Gravity | 1.335 |
| Grade | ACS |
| Physical Form | Solid |
| Show More |