Learn More
Urea, 99%, ACS reagent
CAS: 57-13-6 | CH4N2O | 60.06 g/mol
Supplier: Thermo Scientific Chemicals 424581000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Urea | |
| >132°C | |
| White | |
| Authentic | |
| Plastic bottle | |
| H2NCONH2 | |
| 100 g | |
| 01,1262; 02,455 | |
| carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
| XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
| urea | |
| 1176 | |
| 60.06 | |
| 99% | |
| 0.01% max |
| 57-13-6 | |
| 131.0°C to 135.0°C | |
| 0.01% max. | |
| 99% | |
| CH4N2O | |
| MFCD00008022 | |
| 03, 42 | |
| 15, 10052 | |
| Solubility in water: 1080g/L (20°C). Other solubilities: soluble in alcohol,benzene,concentrated hcl,practically insoluble in chloroform and ether,soluble in methanol: 16.6g/100 mL,soluble in ethanol: 5g/100 mL | |
| C(=O)(N)N | |
| 60.06 | |
| CHEBI:48376 | |
| 1.25mmHg at 25°C | |
| ACS Reagent | |
| Fine Crystals |
Chemical Identifiers
| 57-13-6 | |
| XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
| 1176 | |
| urea |
| MFCD00008022 | |
| carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
| CHEBI:48376 | |
| C(=O)(N)N |
Safety and Handling
GHS H Statement:
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 200-315-5
RTECSNumber : YR6250000
TSCA : TSCA
RUO – Research Use Only