missing translation for 'onlineSavingsMsg'
Learn More

Urea, 99%, ACS reagent

Catalog No. AC424585000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424585000 500 g Plastic bottle
AC424581000 100 g Plastic bottle
AC424580025 2.5 kg Plastic bottle
AC424580100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC424585000 Supplier Thermo Scientific Chemicals Supplier No. 424585000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57-13-6 | CH4N2O | 60.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57-13-6
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Decomposition Information >132°C
Melting Point 131.0°C to 135.0°C
Color White
Ignition Residue 0.01% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Molecular Formula CH4N2O
Linear Formula H2NCONH2
Quantity 500 g
Beilstein 03, 42
Fieser 01,1262; 02,455
Merck Index 15, 10052
Solubility Information 0.01% max. Insoluble Matter
Molecular Weight (g/mol) 60.056
Vapor Pressure 1.25mmHg at 25°C
Formula Weight 60.06
Percent Purity 99%
Grade ACS Reagent
Physical Form Fine Crystals
Chemical Name or Material Urea
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-315-5
RTECSNumber YR6250000
TSCA TSCA

RUO – Research Use Only