missing translation for 'onlineSavingsMsg'
Learn More

Urea, ≥99.5%, Molecular Biology Grade, Ultrapure

Catalog No. AAJ75826A7
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ75826A7 5 kg Plastic bottle
AAJ75826A1 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ75826A7 Supplier Thermo Fisher Scientific Supplier No. J75826A7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chaotropic agent used in denaturing polyacrylamide gel electrophoresis. When 8M urea is not sufficiently denaturing, 40% formamide may be included. | CAS: 57-13-6 | CH4N2O | 60.056 g/mol

  • Chaotropic agent used in denaturing polyacrylamide gel electrophoresis.
  • When 8M urea is not sufficiently denaturing, 40% formamide may be included.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 132°C to 135°C
Density 1.335
Packaging Plastic bottle
Linear Formula NH2CONH2
Quantity 5 kg
Beilstein 635724
Specific Gravity 1.335
Formula Weight 60.06
Percent Purity ≥99.5%
Grade Molecular Biology
Physical Form Crystalline Powder
Chemical Name or Material Urea
Show More Show Less
EINECSNumber 200-315-5
RTECSNumber YR6250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only