missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Absorbance | (8M in H2O), (8 M in H2O) at 0nm, 0.05 max. at 260nm, 0.05 max. at 280nm |
| Melting Point | 131.0°C to 135.0°C |
| Color | White |
| Assay Percent Range | 99.5% min |
| Packaging | Glass Bottle |
| Linear Formula | H2NCONH2 |
| Quantity | 1 kg |
| Beilstein | 03, 42 |
| Fieser | 01,1262; 02,455 |
| Merck Index | 15, 10052 |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement: Causes skin irritation. |
| Precautionary Statement | GHS P Statement: IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 200-315-5 |
RUO – Research Use Only