missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free

Catalog No. AC327380010
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327380010 1 kg Glass Bottle
AC327380050 5 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC327380010 Supplier Thermo Scientific Chemicals Supplier No. 327380010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 57-13-6 | CH4N2O | 60.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Absorbance (8M in H2O), (8 M in H2O) at 0nm, 0.05 max. at 260nm, 0.05 max. at 280nm
Melting Point 131.0°C to 135.0°C
Color White
Assay Percent Range 99.5% min
Packaging Glass Bottle
Linear Formula H2NCONH2
Quantity 1 kg
Beilstein 03, 42
Fieser 01,1262; 02,455
Merck Index 15, 10052
Solubility Information Solubility in water: 1080g/L (20°C). Other solubilities: soluble in alcohol, benzene, concentrated hcl, practically insoluble in chloroform and ether, soluble in methanol: 16.6 g/100 mL, soluble in ethanol: 5 g/100 mL
Formula Weight 60.06
Percent Purity 99.5%
Grade Molecular Biology
Physical Form Solid
Chemical Name or Material Urea, DNAse, RNAse and Protease Free
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-315-5

RUO – Research Use Only