missing translation for 'onlineSavingsMsg'
Learn More

Urea, 99.3+%

Catalog No. AA36429A4
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36429A4 2.5 kg
AA3642922 100 g
AA3642936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA36429A4 Supplier Thermo Scientific Chemicals Supplier No. 036429A4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57-13-6 | CH4N2O | 60.056 g/mol

Urea has important use as a fertilizer and feed supplement. Used for the preparation of plastics and drugs. Urea and its derivatives plays an important role in the cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 132°C to 135°C
Color White
Density 1.335 g/mL
pH 9.2 to 9.5
Odor Ammonia-like
Linear Formula NH2CONH2
Quantity 2.5 kg
Beilstein 635724
Merck Index 14,9867
Solubility Information Soluble in water,glycerol and ethanol.
Formula Weight 60.06
Percent Purity ≥99.3%
Physical Form Crystalline Solid
Chemical Name or Material Urea
Show More Show Less
EINECSNumber 200-315-5
RTECSNumber YR6250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only