missing translation for 'onlineSavingsMsg'
Learn More

Urea, ≥98%, Molecular Biology Reagent Grade, MP Biomedicals™

Catalog No. ICN19485780
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19485780 100 g
ICN19485790 500 g
ICN19485791 1 kg
ICN19485794 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19485780 Supplier MP Biomedicals Inc Supplier No. 0219485780

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Color White
Density 1.335g/mL at 25°C (Lit.)
Assay Percent Range ≥98%
Quantity 100 g
Formula Weight 60.06
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material Urea
EINECSNumber 200-315-5
Recommended Storage Store at Room Temperature (15 to 30°C).