missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Urea
Description
Urea
Specifications
Specifications
| CAS | 57-13-6 |
| Melting Point | 132°C to 135°C |
| Density | 1.335 g/mL at 25°C |
| Molecular Formula | CH4N2O |
| Linear Formula | NH2CONH2 |
| MDL Number | MFCD00008022 |
| Quantity | 5 kg |
| Synonym | Carbamide; Carbonyldiamide |
| Molecular Weight (g/mol) | 60.06 g/mol |