missing translation for 'onlineSavingsMsg'
Learn More

Uranine, TCI America™

Catalog No. F009625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F009625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F009625G Supplier TCI America Supplier No. F009625G
Only null left

Chemical Identifiers

CAS 518-47-8
Molecular Formula C20H10Na2O5
Molecular Weight (g/mol) 376.28
MDL Number MFCD00167039
InChI Key NJDNXYGOVLYJHP-UHFFFAOYSA-L
Synonym Fluorescein Sodium Salt
IUPAC Name disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12

Specifications

Formula Weight 376.27
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Uranine
EINECSNumber (5)-0671&(5)-0673&(5)-1416
RTECSNumber LM5425000
TSCA Yes