missing translation for 'onlineSavingsMsg'
Learn More

Uracil 98.0+%, TCI America™

Catalog No. p-7137841
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U001325G 25 g
U0013100G 100 g
U0013500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. U001325G Supplier TCI America Supplier No. U001325G
Only null left

Chemical Identifiers

CAS 66-22-8
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00006016
InChI Key ISAKRJDGNUQOIC-UHFFFAOYSA-N
Synonym uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x
PubChem CID 1174
ChEBI CHEBI:17568
IUPAC Name 1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES O=C1NC=CC(=O)N1

Specifications

Melting Point 335°C
Color White
Quantity 25 g
Formula Weight 112.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Uracil
EINECSNumber (5)-0933
RTECSNumber YQ8650000
TSCA Yes