missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uracil, 99.75%, MP Biomedicals™
$85.64 - $85.64
Chemical Identifiers
| CAS | 66-22-8 |
|---|---|
| Molecular Formula | C4H4N2O2 |
| Molecular Weight (g/mol) | 112.09 |
| MDL Number | MFCD00006016 |
| InChI Key | ISAKRJDGNUQOIC-UHFFFAOYSA-N |
| Synonym | uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x |
| PubChem CID | 1174 |
| ChEBI | CHEBI:17568 |
| IUPAC Name | 1,2,3,4-tetrahydropyrimidine-2,4-dione |
| SMILES | O=C1NC=CC(=O)N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10320425
|
MP Biomedicals Inc
0210320425 |
25 g |
Each for $85.64
|
|
|||||
Chemical Identifiers
| 66-22-8 | |
| 112.09 | |
| ISAKRJDGNUQOIC-UHFFFAOYSA-N | |
| 1174 | |
| 1,2,3,4-tetrahydropyrimidine-2,4-dione |
| C4H4N2O2 | |
| MFCD00006016 | |
| uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x | |
| CHEBI:17568 | |
| O=C1NC=CC(=O)N1 |
Specifications
| 66-22-8 | |
| 99.75% | |
| MFCD00006016 | |
| uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x | |
| O=C1NC=CC(=O)N1 | |
| 112.09 | |
| CHEBI:17568 | |
| Crystalline Powder |
| White | |
| C4H4N2O2 | |
| 25 g | |
| ISAKRJDGNUQOIC-UHFFFAOYSA-N | |
| 1,2,3,4-tetrahydropyrimidine-2,4-dione | |
| 1174 | |
| 112.09 | |
| Uracil |