missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate™ Human Recombinant Protein: Vitronectin

Catalog No. 08126MI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08126MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08126MI Supplier MilliporeSigma™ Supplier No. 08126

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For positive control, cell culture applications

  • Entrez Gene Number: NM_000638.3

Specifications

Concentration 1.5 mg/mL
For Use With (Application) Positive control, Cell Culture
Formulation Liquid in 10mM Sodium Phosphate, 150mM Sodium Chloride, 0.1mM DTT, 0.1mM PMSF and 50% glycerol
pH Range 7.1
Quantity 500 μg
Source N-terminal, GST-tagged, full-length human Vitronectin expressed in E. coli
Product Type Vitronectin
Content And Storage 6 months at -20°C from date of shipment