missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate™ ECL Cell Attachment Matrix

Catalog No. 08110MI
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08110MI 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08110MI Supplier MilliporeSigma™ Supplier No. 08110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for Human Pluripotent Stem Cell Research

  • Derived from EHS mouse tumor
  • Protein determined by Bradford dye binding assay using gamma globulin as standard
  • Frozen solution

Specifications

For Use With (Application) Cell Culture
Formulation 5mg in 5 ml of 0.05 M Tris-HCl; 0.15M NaCl
pH Range 7.4
Quantity 5 mg
Product Type ECL Cell Attachment Matrix
Content And Storage Stable for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing.
Form Frozen