Learn More
MilliporeSigma™ Supelco™ Upchurch Tubing Adapter, 1/4-28 Female, Bore Diam. 0.04 in. (1.0 mm), Tefzel™
Specifications
Specifications
| Packaging Quantity | 1/Pk. |
| Description | Tefzel™ adapter (one-piece design) |
| Material | Tefzel |
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company