Learn More
Idex Valves
Easily switch between two fluid paths without disconnecting the lines
Supplier: Idex UPV101L
Description
Easily switch between two fluid paths without disconnecting the lines.
- Enable connections among multiple fluid systems
Manual valves serve well in applications that require operator control or involve lesser frequency of use or injection of smaller sample volumes. Shut off flow during cleaning, column replacement, or other operations without disconnecting the system.
Specifications
| 0.04 in. | |
| 2.18 in. | |
| 500 psi |
| 6.2 μL | |
| 0.66 in. |