Learn More
Idex Universal Column Couplers
Great for reducing overall flow path volume
Supplier: Idex UPU284
Description
- Couple two devices together
- Lessen volume of flow path
- 10-32 coned port connections
Column Couplers allow zero dead-volume connections between column and column, column and detector, or column and guard column.
Specifications
| Universal Column Coupler | |
| 1.3 μL | |
| Connect Column w/Internal 10-32 Threads to Precolumn or Guard Column w/10-32 Threads | |
| F-200 Derlin Fingertights w/0.007 in. I.D. |