Learn More
Idex Tubing Sleeves DFS Item
Ensure good port connection with capillary tubing
Supplier:  Idex UPF232
 View more versions of this product
Catalog No. 05-700-66
Only null left
Explore available promotions
Description
Description
Ensure good port connection with capillary tubing
  • Maintain concentric connection with minimal dead volume
  • Color coded for easy identification
Sleeves are not permanently attached to a ferrule so they can easily adapt to varying tubing pocket depths minimizing dead volume. Sleeves are designed to be used with 1/16 in. OD, 10-32 threaded fittings to adapt capillary tubing to standard coned ports. Sleeves require a wrench tightened nut to achieve proper sealing and withstand temperatures up to 257°F (125°C) and pressures up to 6000 psi (414 bar).
Specifications
Specifications
Sleeve
0.265 in.
785 μm
PEEK
Natural
1.57 cm
730 to 770 μm O.D. Tubing
Product Suggestions
Product Suggestions