Learn More
Idex Tubing Sleeves
Ensure good port connection with capillary tubing
Supplier: Idex UPF227
Description
Ensure good port connection with capillary tubing
- Maintain concentric connection with minimal dead volume
- Color coded for easy identification
Sleeves are not permanently attached to a ferrule so they can easily adapt to varying tubing pocket depths minimizing dead volume. Sleeves are designed to be used with 1/16 in. OD, 10-32 threaded fittings to adapt capillary tubing to standard coned ports. Sleeves require a wrench tightened nut to achieve proper sealing and withstand temperatures up to 257°F (125°C) and pressures up to 6000 psi (414 bar).
Specifications
| Sleeve | |
| 0.006 to 0.010 in. | |
| 175-215 μm O.D. Tubing |
| Yellow | |
| 230 μm | |
| PEEK |