Learn More
Idex Pressure Relief Valve Tee
Prevent system over pressurization
Supplier: Idex UP P 612
Description
Relief valves protect system components by diverting fluid automatically when inline pressure exceeds the set limit. Use the 100 psi model for general-purpose applications; 5 psi model is perfect for protecting syringe and peristaltic pump systems. To have the pressure relief valve open at a different pressure than 5 or 100 psi, combine a pressure relief tee assembly with a back pressure regulator assembly.Specifications
| 1000 psi |