Learn More
Idex Plastic Tubing Cutter
Spring-loaded guillotine cutters designed specifically for semi-rigid and rigid polymer tubing
Specifications
For Use With Upchurch™ standard polymer tubing cutter
View More Specs
Products 2
Catalog Number Mfr. No. Color Material Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Color Material Product Type Price Quantity & Availability  
0570184
Encompass
View Documents Promotion Details
Idex
UP A 327
Black Plastic Tubing Cutter N/A
Only null left
0570185
Encompass
View Documents Promotion Details
Idex
UPA328
Silver Steel Replacement Blade N/A
Only null left
Description
Description
Spring-loaded guillotine cutters designed specifically for semi-rigid and rigid polymer tubing
  • Built-in guide holes for both 1/16 and 1/8 in. OD tubing
  • Replacement blades included and sold separately
Flat, 90°, burr-free cut
Offers durability, reliability and ease
  • Able to cut through polymers such as PEEK™, FEP, PFA and Tefzel™
  • Specifications
    Specifications
    Upchurch™ standard polymer tubing cutter