Learn More
Idex Open-End Wrenches
Open-end wrenches useful for fixing liquid chromatography equipment
Supplier:  Idex A304
Encompass
Catalog No. 0570047
Only null left
Description
Description
  • Stainless steel
  • Available in three sizes
Specifications
Specifications
Open-end Wrench
0.25 x 0.31 in.
Most Stainless Steel Fittings Used with Liquid Chromatography Equipment
Chrome Plated
0.63 x 0.79 cm
Product Suggestions
Product Suggestions