Learn More
Idex Nut-Extender Tools
Extender tool for flangeless nuts
Supplier: Idex UP P 298
Description
- Material: Delrin™ Acetal
- For use with flangeless nuts
Specifications
| Nut-extender Tool | |
| Acetal | |
| Use to Tighten Any Standard Short or Extra Long Style Flangeless Nuts in Hard-to-Reach Places |