Learn More
Idex LuerTight™ Fittings: Polypropylene Body
Convenience of quick connect and disconnect of a luer connection
Supplier:  Idex UP P 658
Encompass
Catalog No. 05700334
Only null left
Description
Description
Convenience of quick connect and disconnect of a luer connection
  • Manufactured from inert polymers
  • Female Luer x Female 1/4-28 Flat Bottom
  • 0.050 in. thru-hole diameter
  • Manufactured from red PEEK
Specifications
Specifications
Threaded Luer Adapter
PEEK
Red
Female Luer x Female 1/4-28 Flat Bottom
Product Suggestions
Product Suggestions