missing translation for 'onlineSavingsMsg'
Learn More

Idex Threaded Luer Polypropylene Adapter

Convenience of quick connect and disconnect of a luer connection

$10.17

Specifications

Type Luer Adapter
Diameter (English) Inner 0.05 in.
Temperature (Metric) Max 40°C
Material Polypropylene
Color Natural
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
05700147
Encompass
View Documents Promotion Details
Idex
UP P 618/DUP
To Male 1/4-28
Each for $10.17
Only null left
 
05700148
Encompass
View Documents Promotion Details
Idex
UPP 619
To Female 1/4-28
Each for $10.17
Only null left
 
Description

Description

  • Manufactured from inert polymers

Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.

Specifications

Specifications

Luer Adapter
40°C
Natural
0.05 in.
Polypropylene
Videos
SDS
Documents

Documents