Learn More
Idex LuerTight™ Fittings: PEEK/Polypropylene
Convenience of quick connect and disconnect of a luer connection
Supplier: Idex UP P 655
Description
- Manufactured from inert polymers
- Male Luer Lock x Female 1/4-28 Flat Bottom
- Lock hub securely holds female luer adapters in place
- 0.040 in. thru-hole diameter
- Manufactured from PEEK
Specifications
| Luer Adapter | |
| PEEK | |
| Red |