Learn More
Idex Peek Low-Pressure Union Assembly
Make solid connections in low-pressure applications
Supplier: Idex UP P 720
Description
Make solid connections in low-pressure applications
- Manufactured from inert polymers
- 1/4-28 flat-bottom ports
- Max pressure of 1000 psi (69 bar)
- 0.030 in. thru-hole diameter
- 0.41 μL swept volume
Chemical Identifiers
| PEEK |
Specifications
| Union | |
| PEEK | |
| MicroTight Sleeves |
| 2F-125/1P-553, 6-32 | |
| Natural |