Learn More
Idex ZDV Low-Pressure Delrin Acetal Unions DFS Item
Make solid connections in low-pressure applications
$14.23 - $14.23
Specifications
Type Union
Temperature (Metric) Max 60°C
Material Acetal
View More Specs
Products 1
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
05-700-139 DFS Item
View Documents
Idex
P602
Black
Each for $14.23
Only null left
Description
Description
  • Unions do not include fittings—provide your own to match your tubing size
  • Manufactured from inert polymers
  • True ZDV union
  • Acetal body
Specifications
Specifications
Union
60°C
Acetal
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'