Learn More
Idex Low-Pressure Crosses: PEEK
Combine, split, or intersect fluid streams throughout your system
Supplier: Idex UP P723
Description
- Manufactured from inert polymers
- 1/4-28 flat-bottom ports
Use crosses in a variety of low-pressure situations throughout your fluidic system. Select assemblies that come complete with nuts and ferrules or order bodies and provide your own fittings. Each cross comes with handy mounting holes.
Specifications
| Union | |
| 1.25 mm | |
| PEEK |
| 0.05 in. | |
| 1000 psi | |
| Higher Flow Applications with Minimal Flow Path Resistance |