Learn More
Upchurch Scientific™ High-Pressure Unions
Catalog No. 05700248
Encompass
Change view
Click to view available options
Type:
Union
Webbed Union
For Use With (Equipment):
0.020 in. Thru Hole
0.050 in. Thru Hole; External Threads: 3/8-24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
05700248 Webbed Union 0.050 in. Thru Hole; External Threads: 3/8-24
05700225 Union 0.020 in. Thru Hole
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05700248 Supplier Upchurch Scientific™ Supplier No. UP U437
PEEK High-Pressure Unions, Tees, and Crosses hold up to 5000 psi and include FINGERTIGHT III fittings. Fittings work with any 1/16 in. OD tubing.
Specifications
Type Webbed Union
Material Stainless Steel
For Use With (Equipment) 0.050 in. Thru Hole; External Threads: 3/8-24