Learn More
Idex High-Pressure Tees
Combine multiple fluid streams into one under high operating pressures
Supplier:  Idex UP U 466S
Encompass
 View more versions of this product
Catalog No. 29008280
Only null left
Description
Description
  • Securely join fluid pathway components Holds tight connection even through vibration
  • VHP SS tee assembly
  • Max pressure 20,000 psi (1380 bar)
Specifications
Chemical Identifiers
1/16 in. O.D. Tubing
Specifications
Tee
0.5 mm
1/16 in. O.D. Tubing
0.02 in.
PEEK
Product Suggestions
Product Suggestions