Learn More
Idex High-Pressure Tees
Combine multiple fluid streams into one under high operating pressures
Supplier: Idex UPU 430
Description
- Securely join fluid pathway components Holds tight connection even through vibration
- VHP SS tee assembly
- Max pressure 20,000 psi (1380 bar)
Chemical Identifiers
| 1/16 in. O.D. Tubing |
Specifications
| Cross | |
| 0.5 mm | |
| 1/16 in. O.D. Tubing |
| 0.02 in. | |
| Stainless Steel |