Learn More
Idex High-Pressure Crosses
PEEK High-Pressure Unions, Tees, and Crosses hold up to 5000 psi and include FINGERTIGHT III fittings. Fittings work with any 1/16 in. OD tubing.
Supplier: Idex UP P 729/DUP
Specifications
| Cross | |
| 3500 psi | |
| PEEK | |
| 1/16 in. O.D. Tubing |
| 0.02 in. | |
| 0.721 μL | |
| Black |