missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Idex Fingertight Two-Piece Fitting, Long Thread, Natural PEEK, 1/16 in. OD Tubing, 10-32 Coned
Description
Popular threaded coned port configurations support quality high-pressure connections
- Perfect for analytical instruments, in-vitro diagnostics, electronics and other biotechnology applications
Two-piece fittings comprise of a separate nut and ferrule. Save money by replacing only the ferrule. For mass spectrometer electrospray application order ferrule 02019-15.
Specifications
Specifications
| Type | Fingertight Two-Piece Fitting |
| Includes | Ferrule |
| Material | PEEK |
| Color | Natural |
| For Use With (Equipment) | 1/16 in. OD Tubing |